Search
Search
| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677672.3 | XP_005677729.1 | 14 - 15 | 1936 | 111 | |
| XM_005677673.3 | XP_005677730.1 | 13 - 14 | 1900 | 111 | ||
| XM_005677674.3 | XP_005677731.1 | 12 - 13 | 1803 | 111 | ||
| XM_013962444.2 | XP_013817898.1 | 13 - 14 | 1844 | 111 | ||
Set Membership: |
|