Search
Search
| Gene Symbol: SRCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040168.1 | XP_017895657.1 | 14 - 15 | 2556 | 55 | |
| XM_018040169.1 | XP_017895658.1 | 14 - 15 | 2684 | 55 | ||
| XM_018040170.1 | XP_017895659.1 | 14 - 15 | 2450 | 55 | ||
| XM_018040171.1 | XP_017895660.1 | 14 - 15 | 2556 | 55 | ||
Set Membership: |
|