Search
Search
| Gene Symbol: MMP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285765.1 | NP_001272694.1 | 4 - 5 | 719 | 99 | |
| GenBank mRNA | AB010921.1 | - | 4 - 5 | 719 | 99 | |
| KA349457.1 | - | 4 - 5 | 984 | 99 | ||
Set Membership: |
|