Search
Search
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066172.1 | XP_017921661.1 | 3 - 4 | 392 | 63 | |
| XM_018066173.1 | XP_017921662.1 | 3 - 4 | 392 | 63 | ||
| GenBank mRNA | JO612651.2 | - | 398 | 63 | ||
Set Membership: |
|