Search
Search
| Gene Symbol: MYO19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064152.1 | XP_017919641.1 | 13 - 14 | 1617 | 55 | |
| XM_018064153.1 | XP_017919642.1 | 10 - 11 | 1105 | 55 | ||
| XM_018064154.1 | XP_017919643.1 | 13 - 14 | 1557 | 55 | ||
Set Membership: |
|