Search
Search
| Gene Symbol: VNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684767.3 | XP_005684824.2 | 2 - 3 | 356 | 89 | |
| XM_005684768.3 | XP_005684825.2 | 2 - 3 | 356 | 89 | ||
| GenBank mRNA | JO421239.2 | - | 2 - 3 | 379 | 89 | |
| JO430014.2 | - | 6 - 7 | 1310 | 89 | ||
Set Membership: |
|