Search
Search
| Gene Symbol: ELF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314285.1 | NP_001301214.1 | 5 - 6 | 520 | 76 | |
| XM_005690078.3 | XP_005690135.1 | 5 - 6 | 882 | 76 | ||
| XM_005690080.3 | XP_005690137.1 | 5 - 6 | 609 | 76 | ||
| GenBank mRNA | KA331617.1 | - | 2 - 3 | 370 | 76 | |
| KF669900.1 | - | 5 - 6 | 520 | 76 | ||
Set Membership: |
|