Search
Search
| Gene Symbol: GALT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684085.3 | XP_005684142.1 | 5 - 6 | 599 | 79 | |
| XM_005684087.3 | XP_005684144.1 | 5 - 6 | 597 | 79 | ||
| XM_013966145.2 | XP_013821599.1 | 5 - 6 | 621 | 79 | ||
Set Membership: |
|