Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ch03222826_g1
          See other SON GE Assays ›
          Gene Symbol:
          SON
          Gene Name:
          SON DNA binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 380423 - 411661 on Build ARS1
          Species:
          Goat
          Species Specific ID (Flybase ID):
          -
          Assay ID Ch03222826_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SON
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005674676.3 XP_005674733.2 6 - 7 6826 111
          XM_005674677.3 XP_005674734.2 6 - 7 6694 111
          GenBank mRNA JO340813.2 - 5 - 6 3487 111
          JO340814.2 - 4 - 5 3508 111
          JO340815.2 - 4 - 5 3508 111
          JO340817.2 - 5 - 6 3487 111
          JO364925.2 - 6 - 7 1621 111
          JO364928.2 - 6 - 7 1621 111
          JO364931.2 - 6 - 7 1621 111
          JO373656.2 - 2 - 3 218 111
          JO411564.2 - 6 - 7 1620 111
          JO411565.2 - 6 - 7 1620 111
          JO411566.2 - 6 - 7 1620 111
          JO411567.2 - 6 - 7 1620 111
          JO411568.2 - 6 - 7 1673 111
          JO411570.2 - 6 - 7 1673 111
          JO411572.2 - 6 - 7 1673 111
          JO411573.2 - 6 - 7 1620 111
          JO411574.2 - 6 - 7 1673 111
          JO411575.2 - 6 - 7 1620 111
          JO411576.2 - 6 - 7 1673 111
          JO411577.2 - 6 - 7 1673 111
          JO425648.1 - 4 - 5 2585 111
          JO425678.1 - 4 - 5 2564 111
          JO425711.1 - 5 - 6 2543 111
          JO497209.2 - 4 - 5 694 111
          JO497315.2 - 4 - 5 694 111
          JO498566.2 - 4 - 5 694 111
          JO498792.2 - 4 - 5 694 111
          JO555436.2 - 4 - 5 888 111
          JO555438.2 - 4 - 5 888 111
          JO613137.2 - 2 - 3 937 111
          JO613140.2 - 2 - 3 937 111
          JO613141.2 - 2 - 3 937 111
          JO613142.2 - 6 - 7 1685 111
          JO613143.2 - 6 - 7 1632 111
          JO613144.2 - 6 - 7 1685 111
          JO613145.2 - 2 - 3 937 111
          JO613146.2 - 6 - 7 1685 111
          JO613147.2 - 6 - 7 1632 111
          JO613149.2 - 2 - 3 937 111
          JO613150.2 - 2 - 3 937 111
          JO613555.2 - 6 - 7 1632 111
          JO613556.2 - 6 - 7 1632 111
          JO613559.2 - 6 - 7 1632 111
          JO613560.2 - 2 - 3 937 111
          JO613562.2 - 2 - 3 937 111
          JO613563.2 - 6 - 7 1632 111
          JO613566.2 - 6 - 7 1685 111
          JO613567.2 - 6 - 7 1685 111
          JO613568.2 - 6 - 7 1632 111
          JO613569.2 - 6 - 7 1685 111
          JO613570.2 - 6 - 7 1685 111
          JO613572.2 - 6 - 7 1632 111
          JO613573.2 - 6 - 7 1685 111
          JO613574.2 - 2 - 3 937 111
          JO613575.2 - 6 - 7 1685 111
          JO613576.2 - 6 - 7 1632 111
          JO631894.2 - 6 - 7 1149 111
          JO631895.2 - 6 - 7 1096 111
          KA327757.1 - 6 - 7 1650 111

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline