Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ch03222828_g1
          See other SON GE Assays ›
          Gene Symbol:
          SON
          Gene Name:
          SON DNA binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 380423 - 411661 on Build ARS1
          Species:
          Goat
          Species Specific ID (Flybase ID):
          -
          Assay ID Ch03222828_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SON
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005674676.3 XP_005674733.2 8 - 9 7049 92
          XM_005674677.3 XP_005674734.2 8 - 9 6917 92
          GenBank mRNA JO340815.2 - 6 - 7 3731 92
          JO340817.2 - 7 - 8 3710 92
          JO364925.2 - 4 - 5 1398 92
          JO364928.2 - 4 - 5 1398 92
          JO364931.2 - 4 - 5 1398 92
          JO411564.2 - 4 - 5 1397 92
          JO411565.2 - 4 - 5 1397 92
          JO411566.2 - 4 - 5 1397 92
          JO411567.2 - 4 - 5 1397 92
          JO411568.2 - 4 - 5 1450 92
          JO411570.2 - 4 - 5 1450 92
          JO411572.2 - 4 - 5 1450 92
          JO411573.2 - 4 - 5 1397 92
          JO411574.2 - 4 - 5 1450 92
          JO411575.2 - 4 - 5 1397 92
          JO411576.2 - 4 - 5 1450 92
          JO411577.2 - 4 - 5 1450 92
          JO425648.1 - 6 - 7 2808 92
          JO425678.1 - 6 - 7 2787 92
          JO425711.1 - 7 - 8 2766 92
          JO497209.2 - 6 - 7 917 92
          JO497315.2 - 6 - 7 917 92
          JO498566.2 - 6 - 7 917 92
          JO498792.2 - 6 - 7 917 92
          JO555436.2 - 6 - 7 1111 92
          JO555438.2 - 6 - 7 1111 92
          JO613142.2 - 4 - 5 1462 92
          JO613143.2 - 4 - 5 1409 92
          JO613144.2 - 4 - 5 1462 92
          JO613146.2 - 4 - 5 1462 92
          JO613147.2 - 4 - 5 1409 92
          JO613555.2 - 4 - 5 1409 92
          JO613556.2 - 4 - 5 1409 92
          JO613559.2 - 4 - 5 1409 92
          JO613563.2 - 4 - 5 1409 92
          JO613566.2 - 4 - 5 1462 92
          JO613567.2 - 4 - 5 1462 92
          JO613568.2 - 4 - 5 1409 92
          JO613569.2 - 4 - 5 1462 92
          JO613570.2 - 4 - 5 1462 92
          JO613572.2 - 4 - 5 1409 92
          JO613573.2 - 4 - 5 1462 92
          JO613575.2 - 4 - 5 1462 92
          JO613576.2 - 4 - 5 1409 92
          JO631894.2 - 4 - 5 926 92
          JO631895.2 - 4 - 5 873 92
          JT738540.1 - 4 - 5 1349 92
          KA327757.1 - 4 - 5 1427 92

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline