Search
Search
| Gene Symbol: AAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059521.1 | XP_017915010.1 | 8 - 9 | 1102 | 60 | |
| XM_018059525.1 | XP_017915014.1 | 8 - 9 | 1106 | 60 | ||
| GenBank mRNA | JO643483.2 | - | 8 - 9 | 1105 | 60 | |
Set Membership: |
|