Search
Search

SLC43A3| Gene Symbol: SLC43A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059068.1 | XP_017914557.1 | 5 - 6 | 745 | 59 | |
| XM_018059069.1 | XP_017914558.1 | 5 - 6 | 750 | 59 | ||
| XM_018059070.1 | XP_017914559.1 | 5 - 6 | 673 | 59 | ||
| XM_018059071.1 | XP_017914560.1 | 5 - 6 | 672 | 59 | ||
| XM_018059072.1 | XP_017914561.1 | 5 - 6 | 712 | 59 | ||
| GenBank mRNA | JO361028.2 | - | 5 - 6 | 930 | 59 | |
| JO361029.2 | - | 5 - 6 | 930 | 59 | ||
| KA318351.1 | - | 8 - 9 | 1988 | 59 | ||
Set Membership: |
|