Search
Search

SH3GLB1| Gene Symbol: SH3GLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045694.1 | XP_017901183.1 | 6 - 7 | 1005 | 72 | |
| XM_018045695.1 | XP_017901184.1 | 6 - 7 | 1005 | 72 | ||
| XM_018045696.1 | XP_017901185.1 | 7 - 8 | 1092 | 72 | ||
| GenBank mRNA | JO622191.2 | - | 3 - 4 | 492 | 72 | |
Set Membership: |
|