Search
Search
| Gene Symbol: DPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690613.3 | XP_005690670.1 | 2 - 3 | 486 | 64 | |
| GenBank mRNA | JO499248.2 | - | 2 - 3 | 1227 | 64 | |
| JO500657.2 | - | 2 - 3 | 1227 | 64 | ||
Set Membership: |
|