Search
Search

SEZ6L2| Gene Symbol: SEZ6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041025.1 | XP_017896514.1 | 4 - 5 | 1202 | 57 | |
| XM_018041026.1 | XP_017896515.1 | 4 - 5 | 1203 | 57 | ||
| XM_018041027.1 | XP_017896516.1 | 4 - 5 | 1202 | 57 | ||
| XM_018041028.1 | XP_017896517.1 | 4 - 5 | 1203 | 57 | ||
| GenBank mRNA | JO632175.2 | - | 4 - 5 | 1407 | 57 | |
| JO632176.2 | - | 4 - 5 | 1407 | 57 | ||
Set Membership: |
|