Search
Search
| Gene Symbol: IMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675815.3 | XP_005675872.1 | 6 - 7 | 686 | 56 | |
| XM_005675817.3 | XP_005675874.1 | 6 - 7 | 625 | 56 | ||
| GenBank mRNA | JO623515.2 | - | 10 - 11 | 2687 | 56 | |
| KA353214.1 | - | 9 - 10 | 2436 | 56 | ||
Set Membership: |
|