Search
Search
| Gene Symbol: PHTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677866.2 | XP_005677923.1 | 11 - 12 | 1538 | 67 | |
| XM_018045933.1 | XP_017901422.1 | 10 - 11 | 1403 | 67 | ||
| GenBank mRNA | JT686694.1 | - | 12 - 13 | 1640 | 67 | |
Set Membership: |
|