Search
Search

L3MBTL3| Gene Symbol: L3MBTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684735.3 | XP_005684792.1 | 20 - 21 | 2135 | 57 | |
| XM_018053198.1 | XP_017908687.1 | 20 - 21 | 2323 | 57 | ||
| XM_018053199.1 | XP_017908688.1 | 20 - 21 | 2412 | 57 | ||
| XM_018053200.1 | XP_017908689.1 | 20 - 21 | 2320 | 57 | ||
| XM_018053201.1 | XP_017908690.1 | 19 - 20 | 2248 | 57 | ||
| XM_018053202.1 | XP_017908691.1 | 19 - 20 | 2245 | 57 | ||
Set Membership: |
|