Search
Search
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043496.1 | XP_017898985.1 | 3 - 4 | 862 | 80 | |
| GenBank mRNA | JI861834.1 | - | 3 - 4 | 512 | 80 | |
| JT754443.1 | - | 3 - 4 | 520 | 80 | ||
Set Membership: |
|