Search
Search
| Gene Symbol: DSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053037.1 | XP_017908526.1 | 2 - 3 | 564 | 70 | |
| XM_018053038.1 | XP_017908527.1 | 2 - 3 | 564 | 70 | ||
| GenBank mRNA | JT631651.1 | - | 5 - 6 | 2624 | 70 | |
| JT632271.1 | - | 5 - 6 | 2624 | 70 | ||
Set Membership: |
|