Search
Search

SLC44A2| Gene Symbol: SLC44A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051065.1 | XP_017906554.1 | 7 - 8 | 699 | 57 | |
| XM_018051066.1 | XP_017906555.1 | 7 - 8 | 591 | 57 | ||
| XM_018051067.1 | XP_017906556.1 | 7 - 8 | 697 | 57 | ||
| GenBank mRNA | JO367423.2 | - | 7 - 8 | 648 | 57 | |
Set Membership: |
|