Search
Search
| Gene Symbol: ISL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065772.1 | XP_017921261.1 | 4 - 5 | 1289 | 70 | |
| GenBank mRNA | JO364509.2 | - | 2 - 3 | 274 | 70 | |
| JT749356.1 | - | 2 - 3 | 564 | 70 | ||
Set Membership: |
|