Search
Search
| Gene Symbol: MCPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041973.1 | XP_017897462.1 | 11 - 12 | 2187 | 60 | |
| XM_018041974.1 | XP_017897463.1 | 11 - 12 | 2124 | 60 | ||
| XM_018041975.1 | XP_017897464.1 | 8 - 9 | 1918 | 60 | ||
| GenBank mRNA | KA343926.1 | - | 4 - 5 | 332 | 60 | |
Set Membership: |
|