Search
Search
| Gene Symbol: CASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679555.3 | XP_005679612.3 | 7 - 8 | 983 | 56 | |
| XM_018046844.1 | XP_017902333.1 | 7 - 8 | 987 | 56 | ||
| GenBank mRNA | KA309987.1 | - | 4 - 5 | 1712 | 56 | |
Set Membership: |
|