Search
Search
| Gene Symbol: N4BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050581.1 | XP_017906070.1 | 2 - 3 | 628 | 69 | |
| XM_018050582.1 | XP_017906071.1 | 2 - 3 | 571 | 69 | ||
| XM_018050583.1 | XP_017906072.1 | 2 - 3 | 626 | 69 | ||
| GenBank mRNA | JR115780.1 | - | 3 - 4 | 459 | 69 | |
| KA351871.1 | - | 2 - 3 | 595 | 69 | ||
Set Membership: |
|