Search
Search
| Gene Symbol: LTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674720.3 | XP_005674777.2 | 29 - 30 | 5375 | 101 | |
| GenBank mRNA | JO364934.2 | - | 1 - 2 | 109 | 102 | |
| JO434258.2 | - | 7 - 8 | 1305 | 101 | ||
| JO588617.2 | - | 10 - 11 | 1697 | 101 | ||
| JO625812.2 | - | 1 - 2 | 141 | 101 | ||
| JT705976.1 | - | 29 - 30 | 5217 | 101 | ||
| JT717210.1 | - | 7 - 8 | 1310 | 101 | ||
| JT738021.1 | - | 16 - 17 | 2438 | 101 | ||
| KA338687.1 | - | 1 - 2 | 155 | 101 | ||
Set Membership: |
|