Search
Search

NEURL4| Gene Symbol: NEURL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064561.1 | XP_017920050.1 | 26 - 27 | 4572 | 87 | |
| XM_018064562.1 | XP_017920051.1 | 24 - 25 | 3949 | 87 | ||
| XM_018064563.1 | XP_017920052.1 | 23 - 24 | 3882 | 87 | ||
| GenBank mRNA | JO621349.2 | - | 23 - 24 | 4179 | 87 | |
Set Membership: |
|