Search
Search
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679823.3 | XP_005679880.2 | 6 - 7 | 1349 | 68 | |
| XM_018047628.1 | XP_017903117.1 | 7 - 8 | 1400 | 68 | ||
| GenBank mRNA | JO595519.2 | - | 10 - 11 | 1572 | 68 | |
Set Membership: |
|