Search
Search
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679823.3 | XP_005679880.2 | 10 - 11 | 1890 | 112 | |
| XM_005679824.3 | XP_005679881.2 | 9 - 10 | 1641 | 112 | ||
| XM_018047628.1 | XP_017903117.1 | 11 - 12 | 1941 | 112 | ||
| GenBank mRNA | JO595519.2 | - | 6 - 7 | 1031 | 112 | |
Set Membership: |
|