Search
Search

FBXO38| Gene Symbol: FBXO38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683134.3 | XP_005683191.1 | 8 - 9 | 1118 | 75 | |
| XM_005683135.3 | XP_005683192.1 | 9 - 10 | 1144 | 75 | ||
| XM_005683137.2 | XP_005683194.1 | 8 - 9 | 1128 | 75 | ||
| XM_005683138.2 | XP_005683195.1 | 8 - 9 | 1128 | 75 | ||