Search
Search

PPP4R3B| Gene Symbol: PPP4R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686623.2 | XP_005686680.1 | 3 - 4 | 630 | 71 | |
| XM_005686624.3 | XP_005686681.1 | 3 - 4 | 297 | 71 | ||
| GenBank mRNA | JO425415.1 | - | 13 - 14 | 3342 | 71 | |
Set Membership: |
|