Search
Search
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060225.1 | XP_017915714.1 | 6 - 7 | 1201 | 68 | |
| GenBank mRNA | JO365657.2 | - | 2 - 3 | 1073 | 68 | |
| JO500843.2 | - | 1061 | 68 | |||
Set Membership: |
|