Search
Search
| Gene Symbol: MGAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062074.1 | XP_017917563.1 | 12 - 13 | 1925 | 60 | |
| XM_018062080.1 | XP_017917569.1 | 13 - 14 | 2010 | 60 | ||
| XM_018062085.1 | XP_017917574.1 | 11 - 12 | 1819 | 60 | ||
| XM_018062092.1 | XP_017917581.1 | 12 - 13 | 1922 | 60 | ||
| GenBank mRNA | JT694103.1 | - | 9 - 10 | 2033 | 60 | |
Set Membership: |
|