Search
Search
| Gene Symbol: DLG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043862.1 | XP_017899351.1 | 15 - 16 | 2292 | 64 | |
| XM_018043863.1 | XP_017899352.1 | 15 - 16 | 2239 | 64 | ||
| XM_018043864.1 | XP_017899353.1 | 14 - 15 | 2193 | 64 | ||
| XM_018043865.1 | XP_017899354.1 | 9 - 10 | 1251 | 64 | ||
| XM_018043866.1 | XP_017899355.1 | 7 - 8 | 729 | 64 | ||
Set Membership: |
|