Search
Search
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676426.2 | XP_005676483.1 | 2 - 3 | 1210 | 95 | |
| GenBank mRNA | JT671845.1 | - | 6 - 7 | 2066 | 95 | |
| KU041750.1 | - | 2 - 3 | 799 | 95 | ||
Set Membership: |
|