Search
Search
| Gene Symbol: UVRAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059329.1 | XP_017914818.1 | 12 - 13 | 1406 | 137 | |
| XM_018059330.1 | XP_017914819.1 | 13 - 14 | 1289 | 137 | ||
| GenBank mRNA | JO611193.2 | - | 3 - 4 | 3259 | 137 | |
Set Membership: |
|