Search
Search
| Gene Symbol: IDO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314289.1 | NP_001301218.1 | 3 - 4 | 310 | 63 | |
| GenBank mRNA | JO579703.1 | - | 7 - 8 | 1394 | 63 | |
| KF819363.1 | - | 3 - 4 | 310 | 63 | ||
Set Membership: |
|