Search
Search
| Gene Symbol: HSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314344.2 | NP_001301273.1 | 9 - 10 | 1135 | 114 | |
| XM_018058071.1 | XP_017913560.1 | 9 - 10 | 1333 | 114 | ||
| XM_018058072.1 | XP_017913561.1 | 9 - 10 | 1333 | 114 | ||
| GenBank mRNA | JQ990220.1 | - | 9 - 10 | 1138 | 114 | |
Set Membership: |
|