Search
Search
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285539.1 | NP_001272468.1 | 5 - 6 | 573 | 84 | |
| GenBank mRNA | JO517063.2 | - | 2 - 3 | 118 | 84 | |
| X58471.1 | - | 5 - 6 | 545 | 84 | ||
| Z19569.1 | - | 5 - 6 | 573 | 84 | ||
Set Membership: |
|