Search
Search
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287558.1 | NP_001274487.1 | 3 - 4 | 341 | 86 | |
| GenBank mRNA | HQ606485.1 | - | 3 - 4 | 351 | 86 | |
| JO420454.2 | - | 1 - 2 | 122 | 86 | ||
| JQ031286.1 | - | 3 - 4 | 341 | 86 | ||
Set Membership: |
|