Search
Search
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697627.3 | XP_005697684.1 | 7 - 8 | 1353 | 62 | |
| XM_018041047.1 | XP_017896536.1 | 5 - 6 | 1234 | 62 | ||
| GenBank mRNA | JO580252.1 | - | 7 - 8 | 1456 | 62 | |
Set Membership: |
|