Search
Search
| Gene Symbol: KDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681629.3 | XP_005681686.2 | 26 - 27 | 3774 | 85 | |
| GenBank mRNA | JX094508.1 | - | 4 - 5 | 439 | 85 | |
| JX094509.1 | - | 5 - 6 | 474 | 85 | ||
Set Membership: |
|