Search
Search
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049731.1 | XP_017905220.1 | 5 - 6 | 711 | 92 | |
| XM_018049732.1 | XP_017905221.1 | 5 - 6 | 711 | 92 | ||
| XM_018049733.1 | XP_017905222.1 | 5 - 6 | 711 | 92 | ||
| XM_018049734.1 | XP_017905223.1 | 5 - 6 | 711 | 92 | ||
| XM_018049735.1 | XP_017905224.1 | 5 - 6 | 711 | 92 | ||
| GenBank mRNA | JO354890.2 | - | 5 - 6 | 647 | 92 | |
| JO565914.2 | - | 5 - 6 | 638 | 92 | ||
Set Membership: |
|