Search
Search

C17H12orf43| Gene Symbol: C17H12orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691577.3 | XP_005691634.2 | 1 - 2 | 164 | 102 | |
| XM_005691578.3 | XP_005691635.2 | 1 - 2 | 160 | 102 | ||
Set Membership: |
|