Search
Search
| Gene Symbol: ABCA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063921.1 | XP_017919410.1 | 15 - 16 | 2442 | 62 | |
| XM_018063922.1 | XP_017919411.1 | 15 - 16 | 2368 | 62 | ||
| XM_018063923.1 | XP_017919412.1 | 15 - 16 | 2138 | 62 | ||
| XM_018063924.1 | XP_017919413.1 | 15 - 16 | 2100 | 62 | ||
| XM_018063925.1 | XP_017919414.1 | 14 - 15 | 2578 | 62 | ||
Set Membership: |
|