Search
Search

TRIM33| Gene Symbol: TRIM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677991.3 | XP_005678048.2 | 9 - 10 | 1766 | 64 | |
| XM_018045942.1 | XP_017901431.1 | 9 - 10 | 1766 | 64 | ||
| GenBank mRNA | JT630161.1 | - | 14 - 15 | 1516 | 64 | |
| JT686668.1 | - | 13 - 14 | 1596 | 64 | ||
Set Membership: |
|