Search
Search
| Gene Symbol: KIFC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062046.1 | XP_017917535.1 | 6 - 7 | 834 | 54 | |
| XM_018062047.1 | XP_017917536.1 | 6 - 7 | 831 | 54 | ||
| XM_018062048.1 | XP_017917537.1 | 6 - 7 | 834 | 54 | ||
| XM_018062049.1 | XP_017917538.1 | 4 - 5 | 586 | 54 | ||
| XM_018062050.1 | XP_017917539.1 | 4 - 5 | 586 | 54 | ||
| XM_018062052.1 | XP_017917541.1 | 5 - 6 | 690 | 54 | ||
| XM_018062053.1 | XP_017917542.1 | 4 - 5 | 567 | 54 | ||
| XM_018062054.1 | XP_017917543.1 | 4 - 5 | 418 | 54 | ||
| GenBank mRNA | KA310832.1 | - | 3 - 4 | 1175 | 54 | |
Set Membership: |
|