Search
Search
| Gene Symbol: TARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677669.3 | XP_005677726.2 | 6 - 7 | 768 | 58 | |
| XM_013962435.2 | XP_013817889.2 | 6 - 7 | 778 | 58 | ||
| XM_018046035.1 | XP_017901524.1 | 6 - 7 | 874 | 58 | ||
Set Membership: |
|