Search
Search

ARFIP1| Gene Symbol: ARFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691168.3 | XP_005691225.1 | 3 - 4 | 296 | 91 | |
| XM_005691169.3 | XP_005691226.1 | 2 - 3 | 357 | 91 | ||
| XM_005691170.3 | XP_005691227.1 | 2 - 3 | 254 | 91 | ||
| XM_005691171.3 | XP_005691228.1 | 2 - 3 | 213 | 91 | ||
| XM_013970488.2 | XP_013825942.1 | 3 - 4 | 461 | 91 | ||
| XM_013970489.2 | XP_013825943.1 | 3 - 4 | 439 | 91 | ||
| XM_018061602.1 | XP_017917091.1 | 3 - 4 | 437 | 91 | ||
| XM_018061603.1 | XP_017917092.1 | 2 - 3 | 185 | 91 | ||
| XM_018061604.1 | XP_017917093.1 | 3 - 4 | 271 | 91 | ||
| XM_018061605.1 | XP_017917094.1 | 3 - 4 | 321 | 91 | ||
| XM_018061607.1 | XP_017917096.1 | 2 - 3 | 254 | 91 | ||
| XM_018061608.1 | XP_017917097.1 | 3 - 4 | 439 | 91 | ||
Set Membership: |
|