Search
Search

ANKFY1| Gene Symbol: ANKFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693417.3 | XP_005693474.1 | 16 - 17 | 2309 | 57 | |
| XM_005693418.3 | XP_005693475.1 | 16 - 17 | 2306 | 57 | ||
Set Membership: |
|